Skip to Content
Merck
All Photos(1)

Key Documents

117105

Millipore

Adenosine 5′-diphosphate monopotassium salt dihydrate

M.W. 501.3 (dihydrate).

Synonym(s):

ADP, K, Adenosine 5-Diphosphate, Potassium Salt, ADP

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
Inquire


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
1 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C10H14KN5O10P2 · 2H2O
CAS Number:
Molecular Weight:
501.32
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 154

Assay

≥98% dry basis (enzymatic)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

≤0.2% ATP (enzymatic)
≤1% AMP

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[O-]P(OP(OC[C@H]([C@@H](O)[C@H]1O)O[C@H]1N2C3=C(N=C2)C(N)=NC=N3)(O)=O)(O)=O.[K+]

InChI

1S/C10H15N5O10P2.K/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(24-10)1-23-27(21,22)25-26(18,19)20;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H2,11,12,13)(H2,18,19,20);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1

InChI key

ZNCWUOPIJTUALR-MCDZGGTQSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Elevated plasma levels of factor VIII enhance arterial thrombus formation on erosive smooth muscle cell-rich neointima but not normal intima in rabbits.: This study explores the effects of elevated plasma levels of factor VIII on arterial thrombus formation, a crucial aspect in understanding vascular diseases and developing therapeutic interventions, where Adenosine 5 prime-diphosphate (ADP) is used for the analysis of platelet aggregation and plays a pivotal role in thrombosis and hemostasis (Sugita et al., 2024).

  • An energy-conserving reaction in amino acid metabolism catalyzed by arginine synthetase.: This research highlights the biochemical role of ADP in facilitating energy-conserving reactions in amino acid metabolism, particularly through the catalytic action of arginine synthetase, underscoring its potential in metabolic engineering and therapeutic applications (Michimori et al., 2024).

  • Transcriptome analysis of cultured human vascular endothelial cells after gamma-ray irradiation and correlation analysis with ATP, ADP, and adenosine as secondary soluble factors.: ADP is used as a cell culture component to study its impact alongside other nucleotides in modulating cellular responses to radiation, providing insights into cellular defense mechanisms and the potential for developing radioprotective strategies (Fujie et al., 2024).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service