Skip to Content
Merck

Skip To

1171

Adenosine 5′-Diphosphate, Monosodium Salt

Synonym(s):

Adenosine 5′-Diphosphate, Monosodium Salt, ADP, Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H14N5NaO10P2
Molecular Weight:
449.18
NACRES:
NA.51
UNSPSC Code:
12352208
Assay:
≥95% (HPLC)
Form:
solid
Solubility:
water: 50 mg/mL
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 154

Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Formed by the hydrolysis of ATP in tissue by adenosine triphosphatase. Rapidly converted back to ATP via oxidative phosphorylation in mitochondria. εmax = 15.4 X 103 ± 2% M-1 cm-1 (259 nm, in H2O, pH 7.0)

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1191A2754203800
manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

100

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

form

solid

form

solid

form

powder

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white

color

white

color

-

color

white


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters