Skip to Content
Merck
All Photos(2)

Key Documents

1.17085

Millipore

MAS-100 Iso NT® Pressure Test Kit

For testing the tightness of installed MAS-100 Iso line instruments

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,450.00

€1,450.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 PKG
€1,450.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

€1,450.00


Check Cart for Availability

Request more information

material

aluminum gauge

Agency

non-compendial

description

air pressure controller

feature

display

packaging

pkg of 1 kit

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100 Iso®

shipped in

ambient

storage temp.

no temp limit

General description

MAS-100 ISO NT® Pressure Test Kit is an additional accessory designed to be specifically used for MAS-100 ISO NT® air sampler. The MAS 100 Iso family has been specially developed for monitoring and measuring the microbiological contamination at fixed sampling locations in cleanrooms, RABS and isolators. The pressure test kit is used to test the tightness of the instruments which are installed in isolators or clean rooms. The test verifies the tightness of the valves and the internal connections. The pressure test should be performed on each sampling head present, before an adjustment and/or calibration.

Application

MAS-100 ISO NT® Pressure Test Kit is an additional accessory used to test the tightness of the valves and the internal connections. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as in Food, Beverage and Environmental applications.

Legal Information

MAS-100 ISO is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service