Skip to Content
Merck
All Photos(1)

Key Documents

1.17074

Supelco

Spectromelt® C 15 MP

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€119.00

€119.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€119.00

About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

€119.00


Estimated to ship on26 April 2025


Request a Bulk Order

Quality Level

General description

Micropowder grinding and tabletting aid for X-ray fluorescence spectroscopy

Storage and Stability

Storage temperature: no restrictions.

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service