Skip to Content
Merck

Skip To

1.16995

Nitrate Test Strips, reflectometric

3-90 mg/L (NO₃⁻), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Nitrate chemical test strips, Nitrate test strip for instrument reader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 tests
Check Cart for Availability
€ 69,60

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€ 69,60


Check Cart for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nitrate Test, reflectometric, 3-90 mg/L (NO3-), for use with REFLECTOQUANT®

description

quantitative

Quality Segment

specific analyte(s)

nitrate

reaction suitability

reaction type: Griess reaction

measuring range

3-90 mg/L (NO3-)

application(s)

agriculture
aquaculture
beverage testing
drinking water
food testing
ground and surface water
mineral water
seawater
wastewater

compatibility

RQflex® 20, for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 3 - 90 mg/l NO₃⁻ Reflectoquant®
The Reflectoquant Nitrate test strips can be used for quick and easy monitoring of drinking water, process water and wastewater, as well as in aquarium water where limits can quickly be exceeded. They can also be used to determine nitrate in food e.g. potatoes. On the strip nitrate ions are reduced to nitrite ions by a reducing agent, which then react to a red-violet dye that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to check for nitrite interferences or to estimate the nitrate concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for nitrate and nitrite. Further quantitative determination methods can be found with our Spectroquant tests.

Analysis Note

function control: conforms
Assay
- standard 1:
3 - 7 mg/l
- standard 2: 45 - 55 mg/l
- standard 3: 72 - 88 mg/l

Other Notes

Replaces: 16995-1; 16995

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.169711.100201.14773
detection method

reflectometric

detection method

reflectometric

detection method

colorimetric

detection method

photometric (Nitrospectral)

specific analyte(s)

nitrate

specific analyte(s)

nitrate

specific analyte(s)

nitrate

specific analyte(s)

nitrate

reaction suitability

reaction type: Griess reaction

reaction suitability

reaction type: Griess reaction

reaction suitability

reaction type: Griess reaction

reaction suitability

-

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

for use with MQuant® StripScan App

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

measuring range

3-90 mg/L (NO3-)

measuring range

5-225 mg/L (NO3-)

measuring range

10-500 mg/L (NO3-)

measuring range

0.2-20.0 mg/L (NO3-N), 0.89-88.5 mg/L (NO3-)

description

quantitative

description

quantitative

description

semi-quantitative

description

-


Still not finding the right product?

Explore all of our products under Nitrate Test


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters