Skip to Content
Merck
All Photos(1)

Key Documents

1.16954

Supelco

Recalibration Set

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€58.10

€58.10


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
€58.10

About This Item

UNSPSC Code:
41115316
NACRES:
NB.35

€58.10


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

compatibility

RQflex® 20

Quality Level

detection method

reflectometric

storage temp.

no temp limit

General description

The Recalibration Set can be used for the RQflex® 20 reflectometer. The set contains a calibration strip, an internal standard and a barcode strip for calibration. It is used for the initial calibration of an instrument befor the first use (every instrument is delivered with a recalibration set) and for recalibration as stated in the manual.

Linkage

Replaces: 1.16954.0002

Legal Information

RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Reflectometric peroxide testing in food packaging materials with RQflex® 20 reflectometer and Reflectoquant® Test Strips.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service