1.16882
Fractogel® EMD SO3 ⁻ (M) Resin
Synonym(s):
Fractogel® EMD SO₃⁻ (M)
About This Item
Recommended Products
Quality Level
ligand
(Sulfoisobutyl)
description
strong cation exchanger, suspension in 20% ethanol and 150 mM NaCl (40-90 µm)
sterility
sterile (Caustic Stable)
product line
Fractogel®
form
resin
specific analyte(s)
MAbs
parameter
8 bar max. pressure
matrix active group
methacrylate
mean particle size
40-90 μm
capacity
130 mg binding capacity (lysozyme/ml of resin)
transition temp
flash point 35 °C (Does not sustain combustion.)
density
1.43 g/cm3 at 20 °C
bulk density
1000 kg/m3
application(s)
recombinant protein
vaccine development
separation technique
cation exchange
strong cation exchange
storage temp.
2-30°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Increased binding strength at low pH
- High aggregate removal efficiency combined with high dynamic binding capacity
- Lower elution volumes for the highest purity levels
- Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution
Due to the titration behavior, the ion exchange capacity can be used from pH 2 to pH 12. The separation of proteins is based on reversible electrostatic interactions between the surface′s positively charged regions and the support. Proteins are retained efficiently on Fractogel® EMD SO3- (M) when the pH of the buffer is about 1 unit below their isoelectric points (pl).
The strength of the binding depends on the following:
- the buffer system
- pH value of the buffer which determines the surface charge of the protein
- the concentration of the counter ions
- the charge density on the support (protein binding capacity)
Packaging
- 1.16882.0100: Fractogel® EMD SO3- (M) Resin 100ml
- 1.16882.0010: Fractogel® EMD SO3- (M) Resin 10ml
- 1.16880.0500: Fractogel® EMD SO3- (M) Resin 500ml
- 1.16882.5000: Fractogel® EMD SO3- (M) Resin 5L
Analysis Note
Microscopic evaluation: Uniform spherical particles,no agglomerates, no fines
Extractable matter (water): ≤ 0.05 %
Cerium: ≤ 1 µg/g
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 1.0 bar
Particle size (d10): 37 - 45 µm
Particle size (d50): 48 - 60 µm
Particle size (d90): 63 - 77 µm
Colony forming units (TAMC + TYMC): ≤ 100 CFU/ml
Endotoxins: ≤ 1.00 EU/ml
Protein binding capacity (lysozyme): 110 - 150 mg/ml
Functional test (b:a): ≤ 0.4
Functional test (c:d): ≤ 0.45
Functional test: Separation chymotrypsinogen A, cytochrom C and lysozyme
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
95.0 °F
Flash Point(C)
35 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service