Skip to Content
Merck
All Photos(1)

Documents

116818

Sigma-Aldrich

Adenosine 3′,5′-cyclic Monophosphorothioate, 8-Bromo-, Sp-Isomer, Sodium Salt

A potent cell-permeable, metabolically-stable activator of cAMP-dependent protein kinase (PKA).

Synonym(s):

Adenosine 3′,5′-cyclic Monophosphorothioate, 8-Bromo-, Sp-Isomer, Sodium Salt, Sp-8-Br-cAMPS, Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BrN5O5PS · Na
Molecular Weight:
446.15
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 40 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent cell-permeable, metabolically stable activator of cAMP-dependent protein kinase (PKA). Induces the guanine-nucleotide-exchange factor activity of GEFI, also known as Epac (exchange protein directly activated by cAMP), and GEFII towards Rap1 both in vivo and in vitro. Significantly more lipophilic and cell permeable than Sp-cAMPS (Cat. No. 116815). λmax = 264 nm (H2O, pH 7.0); εmax = 17,000 M-1cm-1. Note: 5 µmol = 2.23 mg.
A potent cell-permeable, metabolically-stable activator of cAMP-dependent protein kinase (PKA). Induces guanine nucleotide exchange factor activity of GEFI (also known as Epac) and GEFII towards Rap1 both in vivo and in vitro. Significantly more lipophilic and cell-permeable than Sp-cAMPS (Cat. No. 116815). Note: 5 µmol = 2.23 mg.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKA
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kawasaki, H., et al. 1998. Science 282, 2275.
de Rooij, J., et al. 1998. Nature 396, 474.
Holen, I., et al. 1996. Eur. J. Biochem. 236, 163.
Schaap, P., et al. 1993. J. Biol. Chem.268, 6323.
Yokozaki, H., et al. 1992. Cancer Res.52, 2504.

Legal Information

Sold under license of Patent DE 3,802,865.4 issued to BIOLOG LSI.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service