Skip to Content
Merck
All Photos(1)

Key Documents

1.15930

Sigma-Aldrich

Toluidine blue O (C.I.52040)

for microscopy Certistain®

Synonym(s):

Toluidine blue O (C.I.52040), Toluidine blue O (C.I. 52040) Certistain®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
MDL number:
UNSPSC Code:
41116130
EC Index Number:
202-146-2

Quality Level

form

solid

composition

Dye, ≥85% spectrophotometric assay

pH

2.8 (25 °C in H2O)

solubility

30 g/L

bulk density

410 kg/m3

εmax

1330-1570 at 628-633 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C15H16N3S.ClH/c1-9-6-13-15(8-11(9)16)19-14-7-10(18(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H/q+1;/p-1

InChI key

HNONEKILPDHFOL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Toluidine blue O (C.I. 52040) for microscopy Certistain® is adry dye that is used for the preparation of a staining solution for staining nuclei and for visualization of metachromasia in histological sections of human origin. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (Diaminophenothiazins, spectrophotometrically): ≥ 85 %
Identity
- chloride:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 628 - 633 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): 1330 - 1570
TLC-Test: passes test
Loss on drying (110 °C): ≤ 8 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service