Skip to Content
Merck

Skip To

1.15754

Aluminium oxide

fibers for gooch crucibles

Synonym(s):

Aluminium oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Check Cart for Availability
€ 98,20
1 kg
Check Cart for Availability
€ 1.000,00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NB.51

€ 98,20


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

impurities

≤1.0% Substances extractable by hydrochloric acid

loss

≤0.5% loss on ignition, 900°C

mp

>1650 °C

bulk density

50‑240 kg/m3

storage temp.

2-30°C

SMILES string

[Si](=O)=O.[Si](=O)=O.[Al]65O[Al](O6)O5.[Al]43O[Al](O4)O3.[Al]21O[Al](O2)O1

InChI

1S/6Al.2O2Si.9O/c;;;;;;2*1-3-2;;;;;;;;;

InChI key

KZHJGOXRZJKJNY-UHFFFAOYSA-N

Application


  • Elemental analysis of levitated solid samples by microwave-assisted laser induced breakdown spectroscopy.: This study explores the application of aluminium oxide in enhancing the elemental analysis capabilities of laser induced breakdown spectroscopy, providing a novel approach for high precision chemical analysis in analytical chemistry (Alamri AM et al., 2024).

  • Alternative nano-lithographic tools for shell-isolated nanoparticle enhanced Raman spectroscopy substrates.: The article discusses the utilization of aluminium oxide in the development of advanced nano-lithographic tools, significantly enhancing the performance of Raman spectroscopy for chemical detection and analysis (Srivastava K et al., 2024).

  • Unexpected early loosening of rectangular straight femoral Zweymüller stems with an alumina-reduced surface after total hip arthroplasty-a prospective, double-blind, randomized controlled trial.: This study examines the clinical implications of using aluminium oxide in prosthetic implants, specifically its effect on the longevity and stability of femoral stems in hip arthroplasty (Moret CS et al., 2024).

  • Green Synthesis of Aluminum Oxide Nanoparticles Using Clerodendrum phlomidis and Their Antibacterial, Anti-inflammatory, and Antioxidant Activities.: Investigates the biogenic synthesis of aluminium oxide nanoparticles, highlighting their potential in biomedical applications due to their antibacterial, anti-inflammatory, and antioxidant properties (Thanaraj S et al., 2024).

Analysis Note

Substances extractable by hydrochloric acid: ≤ 1.0 %
Loss on ignition (900 °C): ≤ 0.5 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.010971.01077919241
mp

>1650 °C

mp

2050 °C

mp

2050 °C

mp

-

loss

≤0.5% loss on ignition, 900°C

loss

-

loss

-

loss

-

bulk density

50‑240 kg/m3

bulk density

-

bulk density

-

bulk density

-

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-30°C

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

-

impurities

≤1.0% Substances extractable by hydrochloric acid

impurities

-

impurities

-

impurities

-


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B Inhalation

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

142844-00-6

CAS No.

142844-00-6

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters