1.15546
PVDF double cones for HPLC Columns
for use with stainless steel Capillary connections 0.2 mm ID, 0.5 mm OD
Synonym(s):
Polyvinylidene Fluoride Cones
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
PVDF double cones, for use with stainless steel capillary connections 0.2 mm ID, 0.5 mm OD
technique(s)
HPLC: suitable
Quality Level
compatibility
for use with stainless steel capillary connections 0.2 mm ID, 0.5 mm OD
storage temp.
no temp limit
Related Categories
General description
The package contains 10 PVDF double cones for capillary connection (o.d. 0.5 mm) with knurled nuts Ordering No. 1.15545 or 1.51216.
for stainless steel capillary connections i.d. 0.2 mm, e.d. 0.5 mm
Analysis Note
visual control: passes test
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service