Skip to Content
Merck

1.15355

CYTOCOLOR Cytological standard stain acc. to Szczepanik

for microscopy

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
UNSPSC Code:
41116121
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

IVD

for in vitro diagnostic use

storage temp.

20-25°C

General description

Cytocolor Cytological standard stain acc. Szczepanik - for microscopy kit is a ready-to-use staining kit used for human-medical cell diagnosis and serves the purpose of the cytological investigation of sample material of human origin, for example in human gynecological and clinico-cytological specimen materials, e. g. cervical smears. Cytocolor is a quick staining kit based on the standard staining method according to Szczepanik for use in the analysis of cytology specimens. The cytological standard stainung acc. to Szczepanik is intended for the staining of exfoliative cells in cytological specimens.
This Cytocolor quick staining kit contains modified hematoxylin and polychromatic solutions. Consequently, a cytological sample can be stained in about 3 minutes. First, the cell nuclei are stained with a hematoxylin solution. The polychromatic solution EA 31 contained in the kit is a so-called polychromatic mixture of eosin, light green SF, and vesuvine (Bismarck brown) and serves to enhance the staining reaction in the cytoplasm. Due to their different molecular weights, in relation to the different pore sizes of the cell membrane eosin and light green SF enable a differentiated staining of the structure in the various cycle stages of the cells.
The package is sufficient for up to 1000 applications.
It is an IVD registered kit that is CE certified, thus can be used for clinical diagnostic purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Modified fast Papanicolaou stain

Legal Information

CYTOCOLOR is a trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 1 - STOT SE 3

target_organs

Central nervous system, Eyes,Central nervous system, Kidney, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library