Skip to Content
Merck
All Photos(1)

Key Documents

1.15348

Supelco

Kjeldahl tablets

weight 5 g (tablet)without addition of Se and Hg

Sign Into View Organizational & Contract Pricing

Select a Size

250 TABS
€265.00
1000 TABS
€986.00

€265.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
250 TABS
€265.00
1000 TABS
€986.00

About This Item

UNSPSC Code:
12352402
NACRES:
NB.21

€265.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest more information

form

tablet

Quality Level

specific analyte(s)

nitrogen

technique(s)

photometry: suitable

weight

5 g (tablet)without addition of Se and Hg

storage temp.

no temp limit

General description

(without addition of Se and Hg) 5 g/tablet
Type: Mercury- and selenium-free catalyst Composition:
Na2SO4 47.7%
K2SO4 47.7%
TiO2 2.8%
CuSO4 1.8%

Analysis Note

Appearance: light-grey to light-blue tablets
Application test: conforms

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Total Nitrogen Analysis kit offers a fast, accurate, and cost-effective method for water and wastewater testing.

Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service