Skip to Content
Merck
All Photos(1)

Documents

1.14834

Supelco

Cadmium Test, photometric

photometric, 0.025-1.000 mg/L (Cd), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Cadmium Cell Test, photometric, 0.025-1.000 mg/L (Cd), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests
25 tests

specific analyte(s)

cadmium

measuring range

0.025-1.000 mg/L (Cd)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Cadion derivative)

storage temp.

15-25°C

General description

Method: photometric 0.025 - 1.000 mg/l Cd Spectroquant®

This Spectroquant® Cadmium Cell Test allows the accurate quantification of the cadmium content in aqueous samples.

Method applied: In alkaline solution cadmium ions react with a cadion derivative (cadion = trivial name for 1-(4-nitrophenyl)-3-(4-phenylazophenyl)triazene) to form a red complex which is determined photometrically.

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant® Cell and Reagent Tests are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service