Skip to Content
Merck
All Photos(1)

Key Documents

114825-M

Sigma-Aldrich

Adapalene

A cell-permeable naphthoic acid compound that exhibits selective affinity and agonistic potency toward RARβ and RARγ over RARα.

Synonym(s):

Adapalene, CD271, RAR Agonist V, ADA, Differin, 6-(3-(1-Adamantyl)-4-methoxyphenyl)-2-naphthoic acid, CD 271, Differin, 6-(3-(1-Adamantyl)-4-methoxyphenyl)-2-naphthoic acid, CD 271, CD271, RAR Agonist V, ADA

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€84.70

€84.70


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€84.70

About This Item

Empirical Formula (Hill Notation):
C28H28O3
CAS Number:
Molecular Weight:
412.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€84.70


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 12 mg/mL

storage temp.

2-8°C

SMILES string

O(C)c1c(cc(cc1)c5cc6c(cc(cc6)C(=O)O)cc5)C32CC4CC(C3)CC(C2)C4

InChI

1S/C28H28O3/c1-31-26-7-6-23(21-2-3-22-12-24(27(29)30)5-4-20(22)11-21)13-25(26)28-14-17-8-18(15-28)10-19(9-17)16-28/h2-7,11-13,17-19H,8-10,14-16H2,1H3,(H,29,30)

InChI key

LZCDAPDGXCYOEH-UHFFFAOYSA-N

General description

A cell-permeable naphthoic acid compound that exhibits selective affinity and agonistic potency toward RARβ and RARγ over RARα (Kd = 34, 130, and 1100 nM, respectively; AC50 = 2.3, 9.3, and 22 nM, respectively, in COS-7-based reporter assays) without appreciable affinity toward cytosolic retinoic acid binding proteins CRABP I/II (Kd >>1 µM) or detectable activity toward RXRα (AC50 >1 µM). The biological activity and pharmaceutical efficacy of ADA are generally shown to be superior to those of ATRA/Tretinoin (Cat. No. 554720) both in vitro and in vivo.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Irby, C.E., et al. 2008. J. Adolesc. Health43, 421.
Ocker, M., et al. 2003. Int. J. Cancer107, 453.
Kim, M.J., et al. 2000. J. Invest. Dermatol.114, 349.
Shroot, B., et al. 1997. J. Am. Acad. Dermatol.36, S96.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service