Skip to Content
Merck
All Photos(1)

Key Documents

1.14782

Supelco

Zinc Test Refill pack

for use with MQUANT®

Synonym(s):

Zinc Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing

Select a Size

120 TESTS
€150.00

€150.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
120 TESTS
€150.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€150.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

feature

refill

Quality Level

specific analyte(s)

zinc

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

This refill pack contains all the reagents for the MQuant® test kits 1.14780 and 1.14412 for the colorimetric evaluation of zinc. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical and ecological system.

Method
In sulfuric solution zinc ions react with thiocyanate ions and brilliant green to form a blue-green complex. The zinc concentration is measured semiquantitatively by visual colorimetric evaluation.

Analysis Note

Application testconforms

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service