Skip to Content
Merck
All Photos(2)

Key Documents

1.14756

Supelco

Chromate Test Kit, colorimetric

0.10-10 mg/L (Cr), 0.22-22 mg/L (CrO₄²⁻), MQuant®

Synonym(s):

Chromate Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

300 TESTS
€295.00

€295.00


Estimated to ship on20 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
300 TESTS
€295.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€295.00


Estimated to ship on20 May 2025


Request a Bulk OrderRequest more information

Product Name

Chromate Test, colorimetric, 0.10-10 mg/L (Cr), 0.22-22 mg/L (CrO42-), for use with MQUANT®

specific analyte(s)

chromate

Quality Level

measuring range

0.10-10 mg/L (Cr)
0.22-22 mg/L (CrO42-)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-disk comparator 0.10 - 0.20 - 0.35 - 0.6 - 1.0 - 1.8 - 3.0 - 6.0 - 10 mg/l Cr 0.22 - 0.45 - 0.8 - 1.3 - 2.2 - 4.0 - 6.7 - 13 - 22 mg/l CrO MQuant®
The MQuant® Chromate test (formally MColortest®) measures chromium(VI) present in the sample as chromate or dichromate ions in wastewater and percolating water, groundwater and surface water, industrial water and drinking water.

Method
In weakly phosphoric solution chromium(VI) ions react with diphenylcarbazide to form chromium(III) and diphenylcarbazone, which form a red-violet complex. The chromate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
The test measures chromium(VI) present in the sample as chromate or dichromate ions in wastewater and percolating water, groundwater and surface water, industrial water and drinking water.

Application


  • Environmental Monitoring: Chromate tests are essential for monitoring chromium(VI) levels in water and soil. These tests help in assessing environmental pollution caused by industrial discharge and are crucial for ensuring compliance with environmental regulations. Recent research has focused on the simultaneous extraction of phosphate and chromate using nano mixed (Al-Fe-Zr) oxide adsorbents, demonstrating enhanced sensitivity and accuracy in detecting chromate contamination in water (Pala et al., International Journal of Environmental Analytical Chemistry, 2021).



Analysis Note

Application test: conforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service