1.14564
Sulfate Test, photometric
photometric, 100-1000 mg/L (SO42-), Spectroquant®
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
Sulfate Cell Test, photometric, 100-1000 mg/L (SO42-), Spectroquant®
product line
Spectroquant®
Quality Level
usage
sufficient for 25 tests
specific analyte(s)
sulfate
measuring range
100-1000 mg/L (SO42-)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (Barium sulfate, turbidimetric)
storage temp.
15-25°C
General description
The method is analogous to EPA 375.4, APHA 4500-SO4 2- E, and ASTM D516-11.
This Spectroquant® Sulfate Cell Test allows the accurate quantification of the sulfate content in aqueous samples .
Method applied: Sulfate ions react with barium ions to form slightly soluble barium sulfate. The resulting turbidity is measured photometrically (turbidimetric method).
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Met. Corr. 1
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Photometric or turbidimetric determination subsequent to water extraction/fusion melting
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service