Skip to Content
Merck
All Photos(1)

Key Documents

1.14242

Millipore

Potassium tetrathionate

extra pure, suitable for microbiology

Synonym(s):

Potassium tetrathionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2O6S4
CAS Number:
Molecular Weight:
302.45
MDL number:
UNSPSC Code:
41106212
EC Index Number:
237-702-3

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

solubility

232 g/L

density

2.296 g/cm3

bulk density

770 kg/m3

application(s)

microbiology

storage temp.

2-8°C

InChI

1S/2K.H2O6S4/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

UVTKHPSJNFFIDG-UHFFFAOYSA-L

Analysis Note

Assay (acidimetric): ≥ 80 %
Suitability for microbiology: passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service