Skip to Content
Merck
All Photos(1)

Key Documents

1.13885

Millipore

Yeast Extract for Biotechnology and Microbiology

Prime, for Biotechnology and Microbiology

Synonym(s):

Yeast extract, Extract of yeast, Yeast extract powder

Sign Into View Organizational & Contract Pricing

Select a Size

10 KG
€301.00
25 KG
€775.00

€301.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
10 KG
€301.00
25 KG
€775.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
232-387-9
NACRES:
NA.74

€301.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Product Name

Yeast extract, media component, suitable for microbiology

biological source

yeast

Quality Level

form

solid

autoignition temp.

>200 °C

packaging

pkg of 10 kg
pkg of 25 kg

color

beige

pH

7.0 (20 °C, 10 g/L in H2O)

solubility

410 g/L

density

0.5 g/cm3 at 20 °C

bulk density

490 kg/m3

application(s)

microbiology

storage temp.

2-30°C

General description

Yeast Extract powder is manufactured from selected strains of Saccharomyces cerevisiae. It is manufactured under controlled conditions to retain its vitamin content and other nutritive values such as peptides, free amino acids, nucleic acids, minerals, trace elements, and phosphorous compounds. It is rich in vitamins and is often used to supply these factors in culture media at a concentration of 0.3 - 0.5%.

Application

Yeast extract is used as a bacteriological media component for the cultivation of a variety of microorganisms and molecular genetics applications. It is also suitable for use as a nutritional source in microbial media.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service