Skip to Content
Merck
All Photos(1)

Key Documents

1.12994

Millipore

Florisil® (0.150-0.250 mm)

for residue analysis

Synonym(s):

Florisil® (0.150-0.250 mm)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€90.00
1 KG
€706.00

€90.00


Estimated to ship on24 May 2025



Select a Size

Change View
100 G
€90.00
1 KG
€706.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
215-681-1

€90.00


Estimated to ship on24 May 2025


Quality Level

pH

9.5 (20 °C, 100 g/L in H2O, slurry)

storage temp.

no temp limit

SMILES string

[Si](=O)([O-])[O-].[Mg+2]

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Analysis Note

Sieve analysis (dry sieving; >0,15 mm): ≥ 80 %
Appearance: passes test
Identity (Si): passes test
Identity (Magnesium): passes test

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service