Skip to Content
Merck

Skip To

1.12463

Amberlite IRA-402 Ion Exchange Resin

chloride form, 15-50 mesh, strongly basic

Synonym(s):

Ion exchanger Amberlite IRA-402

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Check Cart for Availability
€ 111,00

About This Item

CAS Number:
UNSPSC Code:
41115711
NACRES:
NB.21
MDL number:

€ 111,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ion exchanger Amberlite IRA-402, (strongly basic anion exchanger), Cl⁻ form

Quality Segment

technique(s)

HPLC: suitable, LPLC: suitable

matrix

PS-DVB

matrix active group

SAX

particle size

300-1180 μm

density

1.063-1.093 mg/L

bulk density

670 g/L

separation technique

anion exchange

storage temp.

15-25°C

Analysis Note

Description: yellowish moist spherules
Total capacity: ≥ 1.0 mol/l
Particle size (0.3 - 1.18 mm): ≥ 90 %

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CG5022435947060
particle size

300-1180 μm

particle size

100-200 mesh (wet)

particle size

16-50 mesh

particle size

-

matrix active group

SAX

matrix active group

-

matrix active group

sulfonic acid

matrix active group

-

separation technique

anion exchange

separation technique

-

separation technique

cation exchange

separation technique

-

matrix

PS-DVB

matrix

4% cross-linked methacrylate

matrix

styrene-divinylbenzene (gel)

matrix

-

technique(s)

HPLC: suitable, LPLC: suitable

technique(s)

activity assay: suitable

technique(s)

LPLC: suitable

technique(s)

-

storage temp.

15-25°C

storage temp.

-

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under Ion exchanger Amberlite IRA-402


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters