Skip to Content
Merck

Skip To

1.12412

Yttrium oxide 99+

for analysis 99+ EMSURE®

Synonym(s):

Yttrium oxide 99+

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 222,00

About This Item

Empirical Formula (Hill Notation):
O3Y2
CAS Number:
Molecular Weight:
225.81
UNSPSC Code:
12352303
EC Index Number:
215-233-5
NACRES:
NA.21
MDL number:

€ 222,00


Check Cart for Availability
Packaging: HDPE bottle

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

EMSURE®

form

solid

loss

≤1.0% loss on ignition, 1000°C

mp

2410 °C

density

5.0 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[Y+3].[Y+3].[O-2].[O-2].[O-2]

InChI

1S/3O.2Y/q3*-2;2*+3

InChI key

RUDFQVOCFDJEEF-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, calc. on the ignited basis): ≥ 99
Loss on ignition (1000 °C): ≤ 1.0 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.192086346381.12410
form

solid

form

powder

form

nanopowder

form

-

density

5.0 g/cm3 at 20 °C

density

7.2 g/cm3 at 20 °C

density

4.56 g/mL at 25 °C (lit.)

density

-

mp

2410 °C

mp

>200 °C (decomposes)

mp

-

mp

-

product line

EMSURE®

product line

EMSURE®

product line

-

product line

-

storage temp.

no temp limit

storage temp.

2-8°C

storage temp.

-

storage temp.

no temp limit

Quality Level

300

Quality Level

300

Quality Level

100

Quality Level

200


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters