Skip to Content
Merck
All Photos(1)

Key Documents

1.12292

Sigma-Aldrich

Sodium 1-octanesulfonate

for surfactant tests

Synonym(s):

Sodium 1-octanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€61.90
5 G
€109.00
25 G
€322.00

€61.90


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
2 G
€61.90
5 G
€109.00
25 G
€322.00

About This Item

Empirical Formula (Hill Notation):
C8H17NaO3S
CAS Number:
Molecular Weight:
216.27
MDL number:
UNSPSC Code:
12352107
EC Index Number:
226-195-4
NACRES:
NA.22

€61.90


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

form

solid

pH

5.5-7.5 (20 °C, 100 g/L in H2O)

mp

>300 °C

bulk density

345 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCCCC

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Application

  • Improvement of Determination Method of Acarbose Tablets by High-Performance Liquid Chromatography with Diode Array Detector: This study enhances the HPLC methodologies using Sodium 1-octanesulfonate for the analysis of acarbose tablets, emphasizing improved accuracy and reliability in pharmaceutical testing (S XU, 2017).
  • Simultaneous determination of fangchinoline and tetrandrine in Qi-Fang-Xi-Bi-granules by RP-HPLC: Demonstrates the utility of Sodium 1-octanesulfonate in enhancing the chromatographic separation of traditional Chinese medicinal components, providing a basis for better quality control in herbal medicine (XG Lu et al., 2015).
  • Application of achiral–chiral two‐dimensional HPLC for separation of phenylalanine and tryptophan enantiomers: The use of Sodium 1-octanesulfonate facilitated the enantioselective separation of amino acids, critical for the pharmaceutical analysis of dietary supplements (A Lomenova et al., 2021).

Analysis Note

Assay (alkalimetric, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): conforms
pH (100 g/l, water): 5.8 - 7.5
TLC-Test: conforms
Loss on drying (120 °C; 4h; Vacuum): ≤ 2.0 %

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service