Skip to Content
Merck
All Photos(1)

Key Documents

1.12196

Sigma-Aldrich

Indium

granular 99+

Synonym(s):

Indium

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€1,110.00

€1,110.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€1,110.00

About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
EC Index Number:
231-180-0
NACRES:
NA.22

€1,110.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

solid

bp

2000 °C

density

7.31 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): ≥ 99.0 %
Particle size: about 2 - 8 mm
Identity (ICP-OES): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service