Skip to Content
Merck
All Photos(2)

Documents

1.11670

Supelco

Formic acid 98-100%

Suprapur®

Synonym(s):

Formic acid, Methanoic acid, Formylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-579-1
NACRES:
NB.21

grade

for inorganic trace analysis

Quality Level

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

Assay

≥98% (acidimetric)
98-100%

form

liquid

autoignition temp.

1004 °F

expl. lim.

12-38 % (v/v)
57 %

concentration

≤100% in formic acid

technique(s)

mass spectrometry (MS): suitable

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 10 g/L in H2O)

bp

100-101 °C (lit.)
101 °C/1013 hPa

mp

8.2-8.4 °C (lit.)

transition temp

flash point 49.5 °C

solubility

soluble (Water: miscible in all proportions)

density

1.22 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1000 ppb
phosphate, phosphite (as PO43-): ≤100 ppb
sulfate, sulfite (as SO42-): ≤500 ppb

cation traces

Ag: ≤0.5 ppb
Al: ≤5 ppb
As: ≤1 ppb
Au: ≤0.5 ppb
Ba: ≤1 ppb
Be: ≤0.5 ppb
Bi: ≤0.5 ppb
Ca: ≤10 ppb
Cd: ≤0.5 ppb
Co: ≤0.5 ppb
Cr: ≤1 ppb
Cu: ≤1 ppb
Fe: ≤5 ppb
Ga: ≤0.5 ppb
Ge: ≤0.5 ppb
Hg: ≤1 ppb
In: ≤0.5 ppb
K: ≤5 ppb
Li: ≤0.5 ppb
Mg: ≤2 ppb
Mn: ≤0.5 ppb
Mo: ≤0.5 ppb
Na: ≤5 ppb
Ni: ≤1 ppb
Pb: ≤1 ppb
Pd: ≤1 ppb
Pt: ≤1 ppb
Sb: ≤1 ppb
Sn: ≤0.5 ppb
Sr: ≤0.5 ppb
Ti: ≤0.5 ppb
Tl: ≤0.5 ppb
U: ≤0.5 ppb
V: ≤0.5 ppb
Zn: ≤5 ppb
Zr: ≤0.5 ppb

SMILES string

OC=O

storage temp.

2-30°C

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Identification of in vivo metabolites of Citri Sarcodactylis Fructus by UHPLC-Q/Orbitrap HRMS.: This study used advanced chromatographic techniques to identify and analyze metabolites, demonstrating the application of formic acid in enhancing detection sensitivity and accuracy in analytical chemistry (Deng et al., 2023).

  • Phosphorus recovery from aqueous product of hydrothermal carbonization of cow manure.: Research explored the recovery of phosphorus from hydrothermally carbonized cow manure, where formic acid played a crucial role in the chemical analysis and enhancement of recovery processes (Qaramaleki et al., 2023).

  • Identification and quality evaluation of Chinese rice wine using UPLC-PDA-QTOF/MS with dual-column separation.: Demonstrates formic acid′s role in optimizing the chromatographic profiles of complex organic compounds in beverages, thereby aiding in their quality assessment and standardization (Xiang et al., 2023).

Linkage

Replaces: 11670-1; 11670

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service