Skip to Content
Merck
All Photos(2)

Key Documents

1.11609

Sigma-Aldrich

Histosec pastilles

solidification point 56-58°C embedding agent for histology

Synonym(s):

Embedding agents for microscopy, Embedding agent, Histosec

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€42.90
4 X 2.5 KG
€274.00
25 KG
€534.00

€42.90


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€42.90
4 X 2.5 KG
€274.00
25 KG
€534.00

About This Item

UNSPSC Code:
41116124
NACRES:
NA.21

€42.90


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

solid

potency

>3000 mg/kg LD50, skin (Rabbit)

IVD

for in vitro diagnostic use

transition temp

flash point >100 °C

bulk density

400 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Histosec pastilles - solidification point 56-58 °C embedding agent for histology, is an embedding medium for histology of samples of human origin. it is a paraffin enriched with polymers and containing DMSO (dimethyl sulfoxide). Histosec pastilles (without DMSO) (Product number 1.15161) is also available. DMSO (dimethyl sulfoxide) increases the rate of penetration of paraffin and provides additional preservation of the tissue structures. Its melting range is between 56 - 58 °C and, within this range any specimen can be processed without fear of damage irrespective of the method used. In most clinical laboratories the paraffinization is performed with an automated embedding system and is performed directly after the histoprocessing. The Histosec pastilles are well soluble with the Intermedia Xylene (isometic mixture) (Cat .no. 108298) or xylene substitutes such as Neo-Clear(Product number 1.09843).The paraffin is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Solidification point (rotating thermometer method): 56 - 58 °C
Effect of heating (at 60 °C/100 g): clear melt within max. 20 hours
Solubility in xylene (at 20 °C/0.1 g in 5 ml): clear solute within max. 20 hours
Cutting results (4 µm slices): conforms
Solubility of slices in xylene: <50 seconds

Legal Information

HISTOSEC is a trademark of Merck KGaA, Darmstadt, Germany
NEO-CLEAR is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Questions

  1. Does the DMSO or polymers added to paraffin affect usage for immunohistochemistry or IHC.

    1 answer
    1. The same paraffins used for general histology are also employed for embedding tissue for immunohistochemistry or IHC. In clinical laboratory applications, products such as Paraplast, Paraffin Pastilles, Histosec Paraffin Pastilles, or similar products are utilized. These products are generally modified to make the paraffin more suitable for histology or IHC. Plain paraffins are not generally considered suitable for most histology or IHC applications.

      DMSO is a common modifier, although it presents some safety concerns when embedding paraffin blocks. Users may get some paraffin on their fingers during tissue embedding with paraffin. For concerns about the safety of coming in contact with DMSO, there are paraffins such as product 1071642504 that use select polymers to enhance the performance of plain paraffin. The polymers, being of higher molecular weight, are not easily absorbed through the skin, making them safer for use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service