Skip to Content
Merck
All Photos(1)

Key Documents

1.11110

Supelco

Calcium Test Kit, titrimetric

titrimetric, for use with MQUANT®

Sign Into View Organizational & Contract Pricing

Select a Size

170 TESTS
€119.00

€119.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
170 TESTS
€119.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€119.00


Estimated to ship on26 May 2025


Request a Bulk OrderRequest more information

Product Name

Calcium Test, titrimetric, for use with MQUANT®

specific analyte(s)

calcium

Quality Level

compatibility

for use with MQUANT®

detection method

titrimetric

storage temp.

15-25°C

General description

Method: titrimetric with titration pipette MQuant®
This MQuant® Calcium test (formally MColortest®) is suitable for the titrimetric determination using a graduated syringe.

Mehod
Calcium ions react with an indicator to form a red-violet complex compound. The indicator is released from this compound by titration with a solution of ethylenedinitrilotetraacetic acid disodium salt dihydrate (Titriplex® III). At the titration end-point the color changes to blue-violet. The calcium concentration is determined from the consumption of titration solution. When the total hardness is also determined, it is possible to calculate the magnesium content.

The test can be used in
  • Groundwater and surface water
  • Waters from aquaculture
  • Drinking water
  • Mineral water
  • Boiler water
  • Wastewater

This test is not suited for seawater.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what is the refill kit number of 111110 calcium test

    1 answer
    1. Unfortunately, there are no refills available for this kit. It is formulated to perform 170 tests, components are not available separately.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service