Skip to Content
Merck
All Photos(1)

Key Documents

1.10707

Millipore

KF streptococcus agar (base)

for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€269.00

€269.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
500 G
€269.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€269.00


Estimated to ship on29 May 2025


Request more information

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.2 (35 °C, 72 g/L in H2O, after autoclaving)

bulk density

540 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Streptococcus spp.

Analysis Note

Appearance (clearness): clear
Appearance (colour): violet
Solidification behaviour (2 hrs., 45 °C): liquid
pH-value (25 °C): 7.0 - 7.4
Growth (Enterococcus faecalis ATCC 11700): good to very good
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): good to very good
Growth (Enterococcus hirae ATCC 8043 (WDCM 00089)): good to very good
Growth (Streptococcus pyogenes ATCC 12344): none to fair
Growth (Streptococcus agalactiae ATCC 13813): none to fair
Growth (Lactobacillus plantarum ATCC 8014): none to fair
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): none
Growth (Enterobacter cloacae ATCC 13047 (WDCM 00083)): none
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): none
Red colonies (Enterococcus faecalis ATCC 11700): +
Red colonies (Enterococcus faecalis ATCC 19433 (WDCM 00009)): +
Red colonies (Enterococcus hirae ATCC 8043 (WDCM 00089)): +(poor)
Red colonies (Streptococcus pyogenes ATCC 12344): -
Red colonies (Streptococcus agalactiae ATCC 13813): -
Red colonies (Lactobacillus plantarum ATCC 8014): -
Colour change to yellow (Enterococcus faecalis ATCC 11700): +
Colour change to yellow (Enterococcus faecalis ATCC 19433 (WDCM 00009)): +
Colour change to yellow (Enterococcus hirae ATCC 8043 (WDCM 00089)): +
Colour change to yellow (Streptococcus pyogenes ATCC 12344): -
Colour change to yellow (Streptococcus agalactiae ATCC 13813): -
Colour change to yellow (Lactobacillus plantarum ATCC 8014): -
Incubation: 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service