Skip to Content
Merck
All Photos(1)

Key Documents

1.10664

Millipore

Test agar pH 8.0 for the inhibitor test

for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€345.00

€345.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
500 G
€345.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€345.00


Estimated to ship on29 May 2025


Request more information

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.9-8.1 (25 °C, 27.5 g/L in H2O, after autoclaving)

solubility

27.5 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.9 - 8.1
Solidification behaviour (2 hrs., 40 °C): liquid
Inhibition zone diameter/Gentamicin 10 µg (Bacillus subtilis BGA): 32 - 44 mm
Inhibition zone diameter/Gentamycin 30 µg (Bacillus subtilis BGA): 35 - 45 mm
Inhibition zone diameter/Penicillin G 10 U/IE (Bacillus subtilis BGA): 40 - 48 mm
Inhibition zone diameter/Streptomycin 10 µg (Bacillus subtilis BGA): 30 - 36 mm
Growth (Bacillus subtilis BGA): good to very good
Growth (Kocuria rhizophila ATCC 9341): good to very good

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service