Skip to Content
Merck
All Photos(1)

Key Documents

1.10614

Supelco

Potassium carbonate

(special grade LAB)

Sign Into View Organizational & Contract Pricing

Select a Size

25 KG
€720.00

€720.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 KG
€720.00

About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes

€720.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

for analytical purposes

Quality Level

pH

11.5-12.5 (20 °C, 0.01 g/L in H2O)

mp

891 °C (lit.)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service