Skip to Content
Merck
All Photos(1)

Documents

1.10028

Supelco

Tin Test Strips, colorimetric

10-200 mg/L (Sn), MQuant®

Synonym(s):

Tin chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Tin Test, colorimetric, 10-200 mg/L (Sn), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

tin

measuring range

10-200 mg/L (Sn)

detection method

colorimetric

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips and reagent, 10 - 25 - 50 - 100 - 200 mg/L (Sn), MQuant®

The MQuant® Tin test includes test strips and all necessary reagents and can be used to test for tin and tin compounds in electroplating baths, reducing agents, stabilizers, catalysts, disinfectants, fungicides and mordants used in the textile industry and, more especially, in liquid foods. In the test, all tin ions are reduced to tin(II) ions. These react with toluene-3,4-dithiol to form a red dye. The tin concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the tin concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® Tin test.

Application


  • The Emission Test for Qualitative Detection of Tin(II): A Plausible Mechanistic Insight.: This study delves into the emission test for the qualitative detection of tin(II) ions, providing a detailed mechanistic insight. The research highlights the utility of this test in analytical chemistry, showcasing its potential applications in various qualitative analyses (Ghosh, 2024).

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service