Skip to Content
Merck

Skip To

1.09986

Cobalt standard

1000 mg Co, (CoCl2 in H2O) Titrisol®

Synonym(s):

Cobalt Reference Standard for ICP/AAS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 amp
Check Cart for Availability
€ 52,20

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 52,20


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

Titrisol®

form

concentrate

concentration

1000 mg Co in H2O

technique(s)

AAS: suitable, ICP: suitable

pH

4.8 (20 °C in H2O)

density

1.04 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application

Cobalt standard can be used as a calibration standard for thedetermination of cobalt in wet animal feeds using graphite furnace atomicabsorption spectrometry (GF-AAS)

Features and Benefits

  • Titrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.

Analysis Note

1000 mg Co in H2O, prepared with high-purity CoCl2 and water.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardEnvironment

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7646-79-9

CAS No.

7646-79-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters