Skip to Content
Merck
All Photos(2)

Key Documents

1.09957

Supelco

Sodium hydroxide solution

c(NaOH) = 0.5 mol/l (0.5 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
€32.20

€32.20


Estimated to ship on30 May 2025


Request more information

Select a Size

Change View
1 AMP
€32.20

About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

€32.20


Estimated to ship on30 May 2025


Request more information

Product Name

Sodium hydroxide solution, for 1000 ml, c(NaOH) = 0.5 mol/l (0.5 N) Titrisol®

grade

reagent

Quality Level

vapor pressure

3 mmHg ( 37 °C)

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

0.5 M

technique(s)

titration: suitable

pH

14 (20 °C in H2O)

density

1.185 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Recovery of Ce and La from phosphogypsum leachate by adsorption using grape wastes.: This study explores the use of sodium hydroxide solution in enhancing the adsorption properties of grape waste for recovering cerium and lanthanum from industrial waste streams. The research underscores the potential of using agricultural by-products in environmental remediation techniques (da Silva RNF et al., 2023).


  • Metal encapsulation of waste foundry sand stabilized with alkali-activated binder: Batch and column leaching tests.: The application of sodium hydroxide solution in the stabilization of waste foundry sand with an alkali-activated binder demonstrates its efficacy in immobilizing hazardous metals, offering insights into sustainable waste management practices (Ferrazzo ST et al., 2023).


  • Lanthanum and magnesium activated palygorskite for adsorption of phosphate in piggery wastewater.: Highlighting the role of sodium hydroxide solution in activating palygorskite for enhanced phosphate adsorption, this study offers a solution to managing nutrients in wastewater, contributing to more effective wastewater treatment technologies (Zhang X et al., 2023).


  • The Influence of Beverages on Resin Composites: An In Vitro Study.: Investigating the effects of various beverages on dental resin composites, this study incorporates the use of sodium hydroxide solution to simulate clinical settings and assess the durability of dental materials, providing essential insights for dental research and practice (Gradinaru I et al., 2023).


Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service