Skip to Content
Merck

Skip To

1.09934

Lithium standard

1000 mg Li, (LiCl in H2O) Titrisol®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 amp
Check Cart for Availability
€ 59,20
€ 44,40

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 44,40

List Price€ 59,20Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

Titrisol®

form

concentrate

concentration

1000 mg Li in H2O

technique(s)

AAS: suitable, ICP: suitable

pH

5.0 (20 °C in H2O)

density

1.07 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application

Lithium standardcan be used as a calibration standard for the quantification of lithium inwhole blood and urine samples using flame atomic absorption spectrometry(FAAS)

Features and Benefits

  • Titrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.

Analysis Note

1000 mg Li in H2O, prepared with high-purity LiCl and water.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.703291.702238.05660
grade

analytical standard

grade

certified reference material

grade

certified reference material

grade

-

technique(s)

AAS: suitable, ICP: suitable

technique(s)

ICP: suitable

technique(s)

AAS: suitable

technique(s)

-

format

single component solution

format

single component solution

format

single component solution

format

-

product line

Titrisol®

product line

Certipur®

product line

Certipur®

product line

-

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

-

storage temp.

15-25°C

storage temp.

-

storage temp.

15-25°C

storage temp.

2-30°C


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

hcodes



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters