Skip to Content
Merck
All Photos(1)

Key Documents

1.09895

Supelco

Titriplex® solution B

1 ml = 10 mg CaO/l using 100 ml of water, concnetrate for preparing 1000 ml volumetric solution for titration, Titrisol®

Synonym(s):

Alkaline, determination

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
€44.00

€44.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
1 AMP
€44.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

€44.00


Estimated to ship on28 May 2025


Request more information

Product Name

Titriplex® solution B, for the determination of alkaline earth in water (hardness), 1 ml = 10 mg CaO/l (≙ 1 German degrees of hardness) using 100 ml of water for 1000 ml Titrisol®

Quality Level

product line

Titrisol®

form

liquid

pH

7.5 (20 °C in H2O)

density

1.10 g/cm3 at 20 °C

storage temp.

15-25°C

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany
Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service