Skip to Content
Merck
All Photos(1)

Key Documents

1.09771

Supelco

N-(Trimethylsilyl)imidazole

for gas chromatography

Synonym(s):

N-(Trimethylsilyl)imidazole, TSIM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2Si
CAS Number:
Molecular Weight:
140.26
MDL number:
UNSPSC Code:
12352103
EC Index Number:
242-040-3

Quality Level

form

liquid

autoignition temp.

540 °C (DIN 51794)

technique(s)

gas chromatography (GC): suitable

kinematic viscosity

2.5 cSt(25 °C)

bp

212 °C (decomposition)

mp

-42 °C

transition temp

flash point 66 °C (DIN 51755 Part 1)

density

0.955-0.959 g/cm3 at 25 °C

storage temp.

2-30°C

InChI

1S/C6H12N2Si/c1-9(2,3)8-5-4-7-6-8/h4-6H,1-3H3

InChI key

YKFRUJSEPGHZFJ-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 95.0
Identity (IR-spectrum): conforms

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service