Skip to Content
Merck
All Photos(1)

Key Documents

1.09288

Supelco

Copper(II) ethylenediamine solution

DIN 54270, for measuring the viscosity of cellulose

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€517.00

€517.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€517.00

About This Item

UNSPSC Code:
12352402
NACRES:
NA.05

€517.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

form

liquid

pH

12.0-12.5 (20 °C, 10 g/L in H2O)

density

1.10 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Identity (IR-spectrum): passes test
Copper: 0.95 - 1.05 mol/l
Ethylendiamine: 1.90 - 2.10 mol/l
pH-value (1 %; water): 12.0 - 12.5
Density (d 20/4): 1.096 - 1.102

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, good time. When can I see the analysis sheet before ordering?

    1 answer
    1. The testing can be found on the Specification Sheet. This document can be found on the Product Detail Page. Please see the link below:
      https://www.sigmaaldrich.com/specification-sheet/SPEC/MDA/CHEM/1.09288/1092881000

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service