Skip to Content
Merck
All Photos(1)

Key Documents

1.09182

Supelco

Fluorescent indicator F₂₅₄

Synonym(s):

Fluorescent indicator F₂₅₄

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€75.40

€75.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€75.40

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171501
EC Index Number:
271-896-0
NACRES:
NA.21

€75.40


Estimated to ship on28 May 2025


Request a Bulk Order

form

solid

Quality Level

impurities

≤0.01% Fe (Iron)
≤0.3% Water

pH

7.0 ( in H2O, suspension)

mp

1500 °C

solubility

<0.1 g/L

density

4.0 g/cm3 at 20 °C

bulk density

1450 kg/m3

storage temp.

no temp limit

SMILES string

[Zn+2].[Zn+2].[Si]([O-])([O-])([O-])[O-]

InChI

1S/O4Si.2Zn/c1-5(2,3)4;;/q-4;2*+2

InChI key

ZOIVSVWBENBHNT-UHFFFAOYSA-N

Linkage

Replaces: 9182-1; 9182

Analysis Note

Identity: conforms
Fe (Iron): ≤ 0.01 %
Water: ≤ 0.3 %
Application test for DC: conforms

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service