Skip to Content
Merck
All Photos(1)

Key Documents

1.09084

Millipore

Dust-cover for collection head

aluminum cover, suitable for air monitoring

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€186.00

€186.00


Estimated to ship on02 June 2025


Request more information

Select a Size

Change View
1 UNIT
€186.00

About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

€186.00


Estimated to ship on02 June 2025


Request more information

material

aluminum cover

Agency

non-compendial

product line

MAS-100®

feature

autoclavable

packaging

pkg of 1 unit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 Eco®
for use with MAS-100 NT®
for use with MAS-100 VF®

storage temp.

no temp limit

General description

Extra dust cover for collection head protects the perforated lid from damage and dirt. All portable MAS-100® instruments come with a dust cover made from anodized aluminum. The dust cover can be autoclaved and disinfected with all disinfectants compatible to anodized aluminum in order to avoid false positive results in high grade cleanrooms.

Application

Extra dust cover for collection head prevents cross-contamination for the perforated lid and the agar media, when the air sampler is dormant.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG
MAS-100 VF is a registered trademark of MBV AG

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service