Skip to Content
Merck
All Photos(1)

Key Documents

1.08961

Supelco

Refractive index standard kit 2

traceable to NIST SRM, traceable to PTB, n20/D 1.4969

Synonym(s):

RI Calibration Kit for refractometers.

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€432.00

€432.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
€432.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€432.00


Estimated to ship on25 May 2025


Request a Bulk Order

Agency

traceable to NIST SRM
traceable to PTB

Quality Level

storage temp.

15-25°C

General description

Toluene/water, traceable to SRM from NIST and PTB n (20°C)=1,4969 Certipur®

Application

Refractive index standard kit 2 for use as a calibration standard for refractometers.

Features and Benefits

  • Five 8ml ampoules of the standard solution and five 8ml ampoules with water are included in each kit
  • Traceable to recognized national standards, and to units of measurement recognized at the National Institute Standards and Technology (NIST) and Physical Technical Institute (PTB)
  • Solutions that are stable, ready to use, and have long minimum shelf lives 
  • A certificate of analysis (CoA) is included with every standard kit

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 1) What is the unit of measurement used for these standards (Brix units, Specific Gravity etc)? 2) What is the range that the calibration kit covers? 3) Once reconstituted, how long are these materials good for?

    1 answer
    1. The method of analysis is the refractive index which is measured by an Abbé-type refractometer at a wavelength of 589,3nm at 20°C and 25°C. Stable solutions have a minimum shelf life of 3 years, this kit is supplied ready to use.

      Please see the technical sheets below for more information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/218/670/specification-108961-refractive-index-standard-kit-2-20140224.pdf

      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/329/626/technical-data-sheet-refractive-index.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service