Skip to Content
Merck
All Photos(1)

Key Documents

1.08780

Supelco

Zinc

granular for analysis, particle size about 3-8 mm EMSURE® ISO

Synonym(s):

Zinc

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€131.00
1 KG
€263.00

€131.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€131.00
1 KG
€263.00

About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
EC Index Number:
231-175-3
NACRES:
NA.21

€131.00


Estimated to ship on24 May 2025


Request a Bulk Order

Agency

reag. ISO

Quality Level

product line

EMSURE®

Assay

≥99.9% (complexometric)

form

granular

bp

906 °C/1013 hPa

mp

419 °C

density

7.13 g/cm3 at 20 °C

cation traces

As: ≤0.00001%
Cd: ≤0.0005%
Cu: ≤0.001%
Fe: ≤0.02%
Pb: ≤0.005%
Sn: ≤0.005%

storage temp.

2-30°C

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (complexometric): ≥ 99.9 %
As (Arsenic): ≤ 0.00001 %
Cd (Cadmium): ≤ 0.0005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.02 %
Pb (Lead): ≤ 0.005 %
Sn (Tin): ≤ 0.005 %
Corresponds to ISO

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service