Skip to Content
Merck
All Photos(1)

Key Documents

1.08390

Supelco

Diethylenetriaminepentaaceticacid

saturated solution, suitable for titration

Synonym(s):

Diethylenetriaminepentaaceticacid

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€138.00

€138.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
250 G
€138.00

About This Item

Empirical Formula (Hill Notation):
C14H23N3O10
CAS Number:
Molecular Weight:
393.35
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-652-8
NACRES:
NA.21

€138.00


Estimated to ship on28 May 2025


Request more information

Product Name

Diethylenetriaminepentaaceticacid, Diethylenetriaminepentaaceticacid. CAS 67-43-6, pH 2 - 3 (H₂O, 20 °C) (saturated solution).

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

2-3 (20 °C in H2O, saturated solution)

mp

>206 °C (Decomposition)

solubility

4.8 g/L

bulk density

580 kg/m3

storage temp.

15-25°C

SMILES string

N(CCN(CC(=O)O)CC(=O)O)(CCN(CC(=O)O)CC(=O)O)CC(=O)O

InChI

1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)

InChI key

QPCDCPDFJACHGM-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, calc. on anhydrous substance): ≥ 99 %
Identity (IR-spectrum): conforms
pH-value (saturated solution): 2.0 - 3.0
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 10 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
K (Potassium): ≤ 50 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 5 ppm
Na (Sodium): ≤ 500 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Zn (Zinc): ≤ 5 ppm
Sulfated ash: ≤ 0.2 %
Water (according to Karl Fischer): ≤ 0.5 %

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service