Skip to Content
Merck
All Photos(2)

Documents

1.08107

Supelco

Tetrahydrofuran

≥99.9% (GC), suitable for DNA amplification and peptide synthesis (max. 0.005% H₂O)

Synonym(s):

THF, Tetramethylene oxide, Oxolane, Butylene oxide, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-726-8
NACRES:
NA.21

product name

Tetrahydrofuran, for DNA and peptide synthesis (max. 0.005% H₂O)

vapor density

2.5 (vs air)

Quality Level

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

technique(s)

DNA amplification: suitable

impurities

≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.005% Peroxide (as H2O2)
≤0.005% Water

evapn. residue

≤10 mg/L

color

APHA: ≤10

refractive index

n20/D 1.407 (lit.)

pH

7-8 (20 °C, 200 g/L in H2O)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

transition temp

flash point -21.5 °C

density

0.889 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Color: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Peroxide (as H₂O₂): ≤ 0.005 %
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.005 %
The product is stabilized with 2,6-Di-tert-butyl-4-methylphenol (BHT).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service