Skip to Content
Merck
All Photos(1)

Key Documents

1.08033

Supelco

Total Hardness Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Total Hardness Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing

Select a Size

300 TESTS
€87.10

€87.10


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
300 TESTS
€87.10

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€87.10


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

feature

refill

Quality Level

specific analyte(s)

calcium, magnesium

compatibility

for use with MCOLORTEST®

detection method

titrimetric

storage temp.

15-25°C

General description

This refill pack contains all the reagents for the MQuant® test kit 1.08039 for the colorimetric evaluation of total hardness. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical and ecological system.

Method
Calcium and magnesium ions react with an indicator to form a red complex compound. The indicator is released from this compound by titration with a solution of ethylenedinitrilotetraacetic acid disodium salt dihydrate (Titriplex® III). At the titration end-point the color changes to green. The total hardness is determined from the consumption of titration solution.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service