Skip to Content
Merck
All Photos(1)

Documents

1.07836

Supelco

Antimony(III) oxide

for analysis EMSURE®

Synonym(s):

Antimony(III) oxide, di-Antimony trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O3Sb2
CAS Number:
Molecular Weight:
291.52
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-175-0
NACRES:
NA.21

vapor pressure

13.3 hPa ( 660 °C)

Quality Level

product line

EMSURE®

Assay

≥99.0% (bromometric)

form

solid

potency

>34600 mg/kg LD50, oral (Rat)

impurities

≤0.01% Substances insoluble in hydrochloric acid
≤0.05% Substances soluble in water

bp

1550 °C/1013 hPa

mp

656 °C (sublimed)

solubility

2.70 mg/L

density

5.4 g/cm3

bulk density

800‑1300 kg/m3

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.01%

cation traces

As: ≤0.03%
Ca: ≤0.002%
Cu: ≤0.002%
Fe: ≤0.003%
K: ≤0.002%
Na: ≤0.002%
Pb: ≤0.025%

storage temp.

2-30°C

InChI

1S/3O.2Sb/q3*-2;2*+3

InChI key

GHPGOEFPKIHBNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (bromometric): ≥ 99.0 %
Substances insoluble in hydrochloric acid: ≤ 0.01 %
Substances soluble in water: ≤ 0.05 %
Chloride (Cl): ≤ 0.05 %
Sulfate (SO₄): ≤ 0.01 %
As (Arsenic): ≤ 0.03 %
Ca (Calcium): ≤ 0.002 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.003 %
K (Potassium): ≤ 0.002 %
Na (Sodium): ≤ 0.002 %
Pb (Lead): ≤ 0.025 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service