Skip to Content
Merck
All Photos(1)

Key Documents

1.07627

Millipore

Pseudomonas CFC Selective Supplement

for Pseudomonas spp., pkg of 10 vials, for the preparation of Pseudomonas CFC Selective Agar

Synonym(s):

Pseudomonas Selective Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 1 VIALS
€210.00

€210.00


Estimated to ship on25 May 2025


Request more information

Select a Size

Change View
10 X 1 VIALS
€210.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

€210.00


Estimated to ship on25 May 2025


Request more information

description

Pseudomonas CFC/CN agar (base) acc. ISO 13720 and ISO 16266 (1.07620)

Quality Level

form

solid

composition

Cephalothin, 25 mg per vial
Cetrimide, 5 mg per vial
Fucidin, 5 mg per vial

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

pH

6.2-6.8 (20 °C, 26.2 g/L in H2O)

solubility

26.6 g/L

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Pseudomonas spp.

General description

The Pseudomonas CFC Selective Supplement is a mixture of 3 different inhibitors in lyophilized form. Cetrimide, Fucidin, and Cephalothin inhibit the accompanying Gram-positive and Gram-negative flora.

Application

Pseudomonas CFC Selective Supplement is an additive for the preparation of Pseudomonas CFC Selective Agar for the detection and enumeration of Pseudomonas spp. from food and animal feed.

Analysis Note

Recovery on test medium
- Pseudomonas aeruginosa ATCC 27853 (WDCM 00025):
≥ 70 %
- Pseudomonas putida ATCC 12633 (WDCM 00117): ≥ 70 %
- Pseudomonas fluorescens ATCC 13525 (WDCM 00115): ≥ 70 %
- Pseudomonas fragi ATCC 27362: ≥ 70 %
- Pseudomonas fragi ATCC 4973 (WDCM 00116): ≥ 70 %
- Burkholderia cepacia ATCC 17759: ≥ 70 %
- Proteus mirabilis ATCC 14153: ≤ 0.01 %
- Staphylococcus aureus ATCC 25923 (WDCM 00034): ≤ 0.01 %
Colonies
- Pseudomonas aeruginosa ATCC 27853 (WDCM 00025):
greenish
- Pseudomonas putida ATCC 12633 (WDCM 00117): greenish
- Pseudomonas fluorescens ATCC 13525 (WDCM 00115): greenish
- Pseudomonas fragi ATCC 27362: milky
- Pseudomonas fragi ATCC 4973 (WDCM 00116): milky
- Burkholderia cepacia ATCC 17759: milky
Incubation: 44 hrs.; 25 °C; aerobic;
Reference: blood-agar (24-48 hrs.; 30 °C; aerobic).

Supplement tested in Pseudomonas selektiv agar (base) art. no. 1.07620.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service