Skip to Content
Merck
All Photos(1)

Key Documents

1.07533

Supelco

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate

metal indicator Reag. Ph Eur

Synonym(s):

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, PAR

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€342.00

€342.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€342.00

About This Item

Empirical Formula (Hill Notation):
C11H8N3NaO2 · xH2O
CAS Number:
Molecular Weight:
237.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
236-339-8
NACRES:
NB.21

€342.00


Estimated to ship on21 May 2025


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

form

solid

pH

9.8 (20 °C, 10 g/L in H2O)

solubility

38 g/L

bulk density

630 kg/m3

storage temp.

2-30°C

InChI

1S/C11H9N3O2/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11/h1-7,16H,(H,12,14)/b13-9+

InChI key

VLCAILLZPUINNF-UKTHLTGXSA-N

Analysis Note

Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 99
Appearance of solution: passes test
Water (according to Karl Fischer): 7 - 8
Suitability as indicator (for metal titration): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service