Skip to Content
Merck
All Photos(2)

Key Documents

1.07509

Sigma-Aldrich

Pararosaniline (chloride) (C.I. 42500)

for microscopy Certistain®

Synonym(s):

Pararosaniline (chloride) (C.I. 42500)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18ClN3
CAS Number:
Molecular Weight:
323.82
MDL number:
UNSPSC Code:
41116130
EC Index Number:
209-321-2

Quality Level

form

crystalline

mp

268-270 °C (decomposes)

solubility

10 g/L

bulk density

500 kg/m3

εmax

2620-3030 at 545-547 nm in 50% ethanol at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C19H17N3.ClH/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;/h1-12,20H,21-22H2;1H

InChI key

JUQPZRLQQYSMEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pararosaniline (chloride) (C.I. 42500) for microscopy Certistain®, is a solid dye used to prepare the Schiff′s reagent required for the PAS reaction, for the staining of mucopolysaccharides in histological tissue specimens. The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for histology.
As Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service