Skip to Content
Merck
All Photos(1)

Documents

1.07365

Supelco

Lead

foil for analysis about 0.25 mm thick EMSURE®

Synonym(s):

Lead

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
EC Index Number:
231-100-4
NACRES:
NA.21

Quality Level

product line

EMSURE®

form

solid

impurities

≤0.001% Ag (Silver)
≤0.001% Fe (Iron)
≤0.002% Cu (Copper)
≤0.002% Ni (Nickel)
≤0.01% As (Arsenic)
≤0.01% Sb (Antimony)
≤0.020% Sn (Tin)
≤0.05% Bi (Bismuth)

bp

1740 °C/1013 hPa

mp

327 °C

density

11.3 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Analysis Note

Ag (Silver): ≤ 0.001 %
As (Arsenic): ≤ 0.01 %
Bi (Bismuth): ≤ 0.05 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
Ni (Nickel): ≤ 0.002 %
Sb (Antimony): ≤ 0.01 %
Sn (Tin): ≤ 0.020 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service